Skip to Content
Merck

Skip To

1.37165

Spermidine

EMPROVE® EXPERT

Synonym(s):

Spermidine, 1,8-Diamino-4-azaoctane; N-(3-Aminopropyl)-1,4-diaminobutane, 1,8-Diamino-4-azaoctane, N-(3-Aminopropyl)-1,4-diaminobutane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
Please contact Customer Service for Availability
CZK 52,000.00
Please contact Customer Service for Availability
CZK 161,000.00

About This Item

Linear Formula:
NH2(CH2)3NH(CH2)4NH2
CAS Number:
Molecular Weight:
145.25
EC Index Number:
204-689-0
UNSPSC Code:
12352103
Beilstein/REAXYS Number:
1698591
MDL number:

CZK 52,000.00


Please contact Customer Service for Availability

Request a Sample
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

grade

GMO free, GMP

description

Aflatoxin-free, Melamin-free, DNase-free, RNase-free

product line

EMPROVE® EXPERT

technique(s)

in vitro transcription (IVT): suitable

impurities

low in endotoxins

refractive index

n20/D 1.479 (lit.)

particle size

(liquid or solidified mass)

density

0.925 g/mL at 25 °C (lit.)

application(s)

pharma/biopharma processes
pharmaceutical

storage temp.

15-25°C

SMILES string

NCCCCNCCCN

InChI

1S/C7H19N3/c8-4-1-2-6-10-7-3-5-9/h10H,1-9H2

InChI key

ATHGHQPFGPMSJY-UHFFFAOYSA-N

General description

Finding the right critical raw material that matches your quality expectation can be challenging. Especially if you are advancing through clinical phases, changing regulatory requirements can raise several questions.

As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Spermidine has several applications beyond its chemical behavior as a base. The amino groups can interact with the phosphate groups of DNA and RNA and stabilize them. In addition, spermidine has a positive impact on transcription by RNA polymerases and can be found in several buffers. Within RNA bioprocessing, spermidine can be used for purification to bind and precipitate DNA. It may also be considered as adjuvant for mRNA vaccines.

The EMPROVE® Expert quality grade provided with this product is tested for microbial and endotoxin levels and is RNase- and DNase-free. Stability studies according to ICH Q7 are running and stability conclusions will be available via EMPROVE® Suite.

Biochem/physiol Actions

Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA; Stimulates T4 polynucleotide kinase activity.
Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA; may be used for purification of DNA binding proteins. Stimulates T4 polynucleotide kinase activity.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Solutions should be sterile-filtered, not autoclaved, if sterile solution is necessary. Spermidine deaminates with time; solutions should be stored frozen. Prepare new solutions frequently.


Still not finding the right product?

Explore all of our products under Spermidine


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

233.6 °F - closed cup

flash_point_c

112 °C - closed cup



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library