Skip to Content
Merck

Skip To

1.37159

DL-Dithiothreitol

EMPROVE® ESSENTIAL

Synonym(s):

DL-Dithiothreitol, Cleland’s reagent, DTT, threo-1,4-Dimercapto-2,3-butanediol, threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Please contact Customer Service for Availability
CZK 7,930.00
100 g
Please contact Customer Service for Availability
CZK 49,700.00
1000 g
Please contact Customer Service for Availability
CZK 281,500.00
10 kg
Please contact Customer Service for Availability
CZK 1,746,100.00

About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
EC Index Number:
222-468-7
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
1719757
MDL number:

CZK 7,930.00


Please contact Customer Service for Availability

Request a Sample
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

product line

EMPROVE® ESSENTIAL

technique(s)

in vitro transcription (IVT): suitable

mp

41-44 °C (lit.)

application(s)

pharma/biopharma processes

storage temp.

−20°C (−15°C to −25°C)

SMILES string

O[C@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

VHJLVAABSRFDPM-QWWZWVQMSA-N

General description

Our SAFC Pharma® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.
M-Clarity Program
As part of our Emprove® Program, our raw materials are offered with Emprove® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.
Our comprehensive portfolio of downstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides. DTT is effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N′-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and is less toxic than 2-mercaptoethanol. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).
DTT is an excellent reagent chemical used to reduce disulfide bonds commonly found in bioprocessing. DTT is used in buffer formulations to maintain and stabilize free -SH in applications such as protein based biotherapeutics and enzyme stability.

Packaging

1.37159.0010 / 10 g / Amber glass bottle
1.37159.0100 / 100 g / Amber glass bottle
1.37159.1000 / 1 kg / LDPE bag with outer HDPE drum
1.37159.9010 / 10 kg / LDPE bag with outer HDPE drum

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC Pharma is a registered trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D0632D977943815
biological source

synthetic

biological source

-

biological source

-

biological source

-

application(s)

pharma/biopharma processes

application(s)

general analytical

application(s)

advanced drug delivery

application(s)

-

technique(s)

in vitro transcription (IVT): suitable

technique(s)

-

technique(s)

-

technique(s)

-

product line

EMPROVE® ESSENTIAL

product line

-

product line

-

product line

BioUltra

mp

41-44 °C (lit.)

mp

41-44 °C (lit.)

mp

41-44 °C (lit.)

mp

41-44 °C (lit.)

Quality Level

500

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under DL-Dithiothreitol


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library