Skip to Content
Merck

Skip To

132229

Chloride Standard Solution

traceable to SRM from NIST 10.0 mg/l Clˉ in H2O

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
CZK 2,040.00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116116

CZK 2,040.00


Please contact Customer Service for Availability

Request a Custom OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

specific analyte(s)

chloride

technique(s)

photometry: suitable

pH

4.7 ( in H2O)

density

1.00 g/cm3 at 20 °C

application(s)

agriculture
aquaculture
boiler and cooling water
construction materials
disposal drainage water
drinking water
electroplating
food testing
ground and surface water
mineral water
mining
power plants
seawater
wastewater

shipped in

ambient

storage temp.

15-25°C

General description

This Spectroquant® Chloride Standard Solution is a high-quality laboratory solution used for internal quality control and method-controlling purposes.

These Spectroquant® standard solutions perfectly complement our ready-to-use test kits and photometers within the Spectroquant® portfolio and are ideal for validating international norm methods described in e.g., ISO, EN, APHA, and EPA.

Please visit the Reference Material Quality Grade website to find a comprehensive overview of our different grades of standards.

Application

Chloride Standard Solution can be used to check the photometric measurement system (reagents, measurement device, handling) and the mode of working.

Features and Benefits

  • Manufactured and tested in DIN EN ISO 9001:2008 accredited laboratory
  • The solution is pre-diluted to a concentration that falls into the range of our Spectroquant® test kits. This eliminates the need for dilution prior to analysis and hence avoids a common source of error
  • Ready-to-use diluted reference materials (RMs) along with expanded measurement uncertainty ensure absolute precision
  • Accompanied with a certificate of analysis (COA) detailing the certified values, including uncertainty and expiry date
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST) or to United States Pharmacopeia (USP)
  • Reliable and reproducible results

Analysis Note

Concentration: 10.0 ± 0.5 mg/L Clˉ in H2O

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1322301322311.70216
form

liquid

form

solution

form

solution

form

-

application(s)

agriculture
aquaculture
boiler and cooling water
construction materials
disposal drainage water
drinking water
electroplating
food testing
ground and surface water
mineral water
mining
power plants
seawater
wastewater

application(s)

agriculture
aquaculture
boiler and cooling water
construction materials
disposal drainage water
drinking water
electroplating
food testing
ground and surface water
mineral water
mining
power plants
seawater
wastewater

application(s)

agriculture
aquaculture
boiler and cooling water
construction materials
disposal drainage water
drinking water
electroplating
food testing
ground and surface water
mineral water
mining
power plants
seawater
wastewater

application(s)

industrial qc
pharmaceutical

pH

4.7 ( in H2O)

pH

4.7 ( in H2O)

pH

4.7 ( in H2O)

pH

-

specific analyte(s)

chloride

specific analyte(s)

chloride

specific analyte(s)

chloride

specific analyte(s)

-

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

AAS: suitable

density

1.00 g/cm3 at 20 °C

density

1 g/cm3 at 20 °C

density

1 g/cm3 at 20 °C

density

1.033 g/cm3 at 20 °C


Still not finding the right product?

Explore all of our products under Chloride Standard Solution


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Is Chloride stanadrd is ISo 17034 certified?

    1 answer
    1. This product is compared to metrology standards such as NIST, but not to ISO standards. No ISO 17034 alternatives are available for this product.

      Helpful?

Reviews

No rating value

Active Filters