Skip to Content
Merck
All Photos(1)

Key Documents

1.20628

Millipore

GranuCult® Plus m-Green Broth

nonselective for yeasts, nonselective for molds

Synonym(s):

BromocresolGreen Yeast and Mold Broth, M-BCG Broth, Yeast and Mold Broth

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
CZK 3,860.00

CZK 3,860.00


Please contact Customer Service for Availability


Select a Size

Change View
500 G
CZK 3,860.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

CZK 3,860.00


Please contact Customer Service for Availability

Agency

EP 2.6.1
JP 4.06
USP 71

Sterilization Compatibility

autoclavable compatible

form

granular

packaging

pkg of 500 g

technique(s)

microbiological culture: suitable

pH

4.6±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

15-25°C

suitability

nonselective for molds
nonselective for yeasts

General description

GranuCult® Plus m-Green Broth is a complex media designed to cater excellent fungal growth. The acidic pH in the medium inhibits bacterial growth with an exception of acidophilic bacteria.
The media composition includes Peptones, acting as a rich source of nitrogen, carbon, and amino acids. The addition of Yeast extract ensures an abundant supply of vital vitamins, while Dextrose serves as an energy source.
To maintain the perfect equilibrium, Potassium phosphate acts as a reliable buffering agent, stabilizing the medium′s pH levels. This medium also includes Magnesium sulfate, thiamine, and diastase, which provide a range of essential ions, minerals, and nutrients, crucial for robust fungal growth.
Bromocresol green acts as a pH indicator and facilitates visualization of fungal growth in the broth.

Application

GranuCult® Plus m-Green Broth is used for the detection of yeast and molds in soft drinks and other beverage samples.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service