Skip to Content
Merck
All Photos(1)

Documents

1.19853

Supelco

LiChrolut® EN (40 - 120 µm)

suitable for solid phase extraction (SPE)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41115711

technique(s)

HPLC: suitable
solid phase extraction (SPE): suitable

Quality Level

application(s)

environmental (analysis)

storage temp.

no temp limit

General description

Lichrolut columns are designed for reliable and rapid solid phase extraction. This is achieved by using strong but reversible interactions between the analyte and the surface of the stationary phase. A variety of surface modifications allow non-polar extractions (Lichrolut RP18 and RP18e), polar extractions (Lichrolut Si and Lichrolut CN), Cation Exchange extraction (Lichrolut SCX), mixed mode extraction (Lichrolut TSC) and non-polar extractions on a polymer phase (Lichrolut EN) which is especially well suited for the extraction of pesticdes and phenols in water or drugs in body fluids.
for solid phase extraction in environmental analysis

Analysis Note

Appearance: passes test
Extractable matter: ≤ 0.05 %
Capacity (coffeine): 450 - 550 mg/g
Capacity (Diisodecyl phthalate): 400 - 600 mg/g

Legal Information

LICHROLUT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service