Skip to Content
Merck
All Photos(1)

Documents

1.14831

Supelco

Silver Test

photometric, 0.25-3.00 mg/L (Ag), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116

product line

Spectroquant®

Quality Level

specific analyte(s)

silver

measuring range

0.25-3.00 mg/L (Ag)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric

storage temp.

15-25°C

General description

Method: photometric 0.25 - 3.00 mg/l Ag Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service