Skip to Content
Merck

Skip To

1.14552

Chromate Cell Test, photometric

photometric, 0.05-2.00 mg/L (Cr), 0.11-4.46 mg/L (CrO4), Spectroquant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 tests
Please contact Customer Service for Availability
CZK 4,530.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

CZK 4,530.00


Please contact Customer Service for Availability

Request a Custom OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Chromate Cell Test, photometric, 0.05-2.00 mg/L (Cr), 0.11-4.46 mg/L (CrO4), Spectroquant®

Quality Segment

usage

sufficient for 25 tests

specific analyte(s)

chromate

measuring range

0.05-2.00 mg/L (Cr), 0.11-4.46 mg/L (CrO4)

technique(s)

photometry: suitable

application(s)

aquaculture
boiler and cooling water
construction materials
disposal drainage water
electroplating
environmental
ground and surface water
milk and dairy products
seawater
wastewater

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

detection method

photometric (Diphenylcarbazide)

storage temp.

15-25°C

General description

(chromium VI and total chromium) Method: photometric 0.05 - 2.00 mg/l Cr 0.11 - 4.46 mg/l CrO₄ Spectroquant®


The method is analogous to APHA 3500-Cr B and DIN 38405-52.


This Spectroquant® Chromate Cell Test allows the accurate quantification of the chromium(VI) and total chromium concentration in aqueous samples.


Method applied: In weakly phosphoric solution chromium(VI) ions react with diphenylcarbazide to form chromium(III) and diphenylcarbazone which form a red-violet complex that is determined photometrically.


The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.


All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.147581.147301.14561
specific analyte(s)

chromate

specific analyte(s)

chromate

specific analyte(s)

chloride

specific analyte(s)

cyanide

detection method

photometric (Diphenylcarbazide)

detection method

photometric (Diphenylcarbazide)

detection method

photometric

detection method

photometric (Barbituric acid, pyridine-carboxylic acid)

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

measuring range

0.05-2.00 mg/L (Cr), 0.11-4.46 mg/L (CrO4)

measuring range

0.010-3.00 mg/L (Cr), 0.02-6.69 mg/L (CrO4)

measuring range

5-125 mg/L (Cl-)

measuring range

0.010-0.500 mg/L (CN-)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 600 (plus), for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 300 (plus)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters