1.14418
Copper Test Kit, colorimetric
0.3-5 mg/L (Cu), MQuant®
Synonym(s):
Copper Visual Test Kit
Select a Size
CZK 4,940.00
Select a Size
About This Item
CZK 4,940.00
Recommended Products
Product Name
Copper Test, colorimetric, 0.3-5 mg/L (Cu), for use with MCOLORTEST®
specific analyte(s)
copper
Quality Level
measuring range
0.3-5 mg/L (Cu)
compatibility
for use with MCOLORTEST®
detection method
colorimetric
storage temp.
15-25°C
General description
- Groundwater, surface water, and seawater
- Drinking water
- Wastewater and percolating water
- Boiler and boiler feed water, cooling water
- Electroplating wastewater
- Food after appropriate sample pretreatment
- Swimming-pool water
Method
In an ammoniacal medium copper(II) ions react with cuprizone to form a blue complex. The copper concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. By the addition of hydrogen peroxide copper(I) ions can be measured at the same time.
The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Using the refill pack 1.18459 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service