Skip to Content
Merck

Skip To

1.12996

Spectromelt® A 20

for X-ray fluorescence analysis, lithium metaborate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kg
Please contact Customer Service for Availability
CZK 18,500.00

About This Item

Empirical Formula (Hill Notation):
BLiO2
CAS Number:
Molecular Weight:
49.75
UNSPSC Code:
12352302
EC Index Number:
236-631-5
NACRES:
NA.21
MDL number:
Form:
solid
Solubility:
water: soluble 141.2  g/L at 20 °C (68 °F)

CZK 18,500.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

mp

840 °C

solubility

water: soluble 141.2  g/L at 20 °C (68 °F)

density

2.18 g/cm3 at at 20 °C (68 °F)

bulk density

1300 kg/m3

anion traces

chloride (Cl-): ≤50 ppm, phosphate (PO43-): ≤20 ppm, silicate (as SiO2): ≤100 ppm, sulfate (SO42-): ≤50 ppm

cation traces

Ag: ≤5 ppm, Al: ≤10, Ba: ≤5 ppm, Ca: ≤20 ppm, Cd: ≤5 ppm, Co: ≤5 ppm, Cr: ≤5 ppm, Cu: ≤5 ppm, Fe: ≤10 ppm, Ga: ≤5 ppm, K: ≤10 ppm, Mg: ≤5 ppm, Mn: ≤5 ppm, Na: ≤20 ppm, Na: ≤5 ppm, Pb: ≤5 ppm, Sr: ≤5 ppm, Zn: ≤5 ppm

storage temp.

2-30°C

SMILES string

[B].[B].[B].[B].[Li].[Li].O.O.O.O.O.O.O

InChI

1S/4B.2Li.7H2O/h;;;;;;7*1H2

InChI key

DPYKRXYVXDYLEY-UHFFFAOYSA-N

Analysis Note

Chloride (Cl): ≤ 50 ppm
Phosphate (PO₄): ≤ 20 ppm
Silicate (SiO₂): ≤ 100 ppm
Sulfate (SO₄): ≤ 50 ppm
Ag (Silver): ≤ 5 ppm
Al (Aluminium): ≤ 10 ppm
Ba (Barium): ≤ 5 ppm
Ca (Calcium): ≤ 20 ppm
Cd (Cadmium): ≤ 5 ppm
Co (Cobalt): ≤ 5 ppm
Cr (Chromium): ≤ 5 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 20 ppm
Ga (Gallium): ≤ 5 ppm
K (Potassium): ≤ 10 ppm
Mg (Magnesium): ≤ 10 ppm
Mn (Manganese): ≤ 5 ppm
Na (Sodium): ≤ 20 ppm
Ni (Nickel): ≤ 5 ppm
Pb (Lead): ≤ 5 ppm
Sr (Strontium): ≤ 5 ppm
Zn (Zinc): ≤ 5 ppm
Loss on ignition (800 °C): ≤ 1.0 %
Bulk density: about 130

Legal Information

SPECTROMELT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.107831.126301.11802
form

solid

form

solid

form

solid

form

solid

solubility

water: soluble 141.2  g/L at 20 °C (68 °F)

solubility

141.2 g/L

solubility

141.2 g/L, water: 141.2 g/L at 20 °C (68 °F - OECD Test Guideline 105)

solubility

50 g/L

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

anion traces

chloride (Cl-): ≤50 ppm, silicate (as SiO2): ≤100 ppm, phosphate (PO43-): ≤20 ppm, sulfate (SO42-): ≤50 ppm

anion traces

chloride (Cl-): ≤50 ppm, phosphate (PO43-): ≤20 ppm, silicate (as SiO2): ≤100 ppm, sulfate (SO42-): ≤50 ppm

anion traces

chloride (Cl-): ≤50 ppm, phosphate (PO43-): ≤20 ppm, silicate (as SiO2): ≤100 ppm, sulfate (SO42-): ≤50 ppm

anion traces

chloride (Cl-): ≤50 ppm, phosphate (PO43-): ≤20 ppm, silicate (as SiO2): ≤100 ppm, sulfate (SO42-): ≤50 ppm

cation traces

Ag: ≤5 ppm, Ba: ≤5 ppm, Cd: ≤5 ppm, Cr: ≤5 ppm, Fe: ≤10 ppm, K: ≤10 ppm, Mn: ≤5 ppm, Na: ≤5 ppm, Sr: ≤5 ppm, Ca: ≤20 ppm, Cu: ≤5 ppm, Mg: ≤5 ppm, Pb: ≤5 ppm, Al: ≤10, Ga: ≤5 ppm, Zn: ≤5 ppm, Co: ≤5 ppm, Na: ≤20 ppm

cation traces

Ag: ≤5 ppm, Al: ≤10, Ba: ≤5 ppm, Ca: ≤20 ppm, Cd: ≤5 ppm, Co: ≤5 ppm, Cr: ≤5 ppm, Cu: ≤5 ppm, Fe: ≤10 ppm, Ga: ≤5 ppm, K: ≤10 ppm, Mg: ≤5 ppm, Mn: ≤5 ppm, Na: ≤20 ppm, Ni: ≤5 ppm, Pb: ≤5 ppm, Sr: ≤5 ppm, Zn: ≤5 ppm

cation traces

Ag: ≤5 ppm, Al: ≤10 ppm, Ba: ≤5 ppm, Ca: ≤20 ppm, Cd: ≤5 ppm, Co: ≤5 ppm, Cr: ≤5 ppm, Cu: ≤5 ppm, Fe: ≤10 ppm, Ga: ≤5 ppm, K: ≤10 ppm, Mg: ≤5 ppm, Mn: ≤5 ppm, Na: ≤20 ppm, Ni: ≤5 ppm, Pb: ≤5 ppm, Sr: ≤5 ppm, Zn: ≤5 ppm

cation traces

Ag: ≤5 ppm, Al: ≤10 ppm, Ba: ≤5 ppm, Ca: ≤20 ppm, Cd: ≤5 ppm, Co: ≤5 ppm, Cr: ≤5 ppm, Cu: ≤5 ppm, Fe: ≤10 ppm, Ga: ≤5 ppm, K: ≤10 ppm, Mg: ≤5 ppm, Mn: ≤5 ppm, Na: ≤20 ppm, Ni: ≤5 ppm, Pb: ≤5 ppm, Sr: ≤5 ppm, Zn: ≤5 ppm

mp

840 °C

mp

930 °C

mp

917 °C ((1683 °F))

mp

-


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters