Skip to Content
Merck
All Photos(1)

Key Documents

1.12406

Sigma-Aldrich

Tungsten

fine powder 99+

Synonym(s):

Tungsten

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
MDL number:
UNSPSC Code:
12141747
EC Index Number:
231-143-9
NACRES:
NA.22

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

mp

3370 °C

bulk density

4000 kg/m3

storage temp.

no temp limit

InChI

1S/W

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

Analysis Note

Identity (elemental analysis): passes test
Assay (gravimetrically): ≥ 99.0 %
Particle size (< 20 µm): ≥ 80 %

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service