Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 g | Please contact Customer Service for Availability | CZK 5,140.00 CZK 3,855.00 |
About This Item
Empirical Formula (Hill Notation):
C8H17NaO4S
CAS Number:
Molecular Weight:
232.27
UNSPSC Code:
12352107
EC Index Number:
205-535-5
NACRES:
NA.22
MDL number:
CZK 3,855.00
List PriceCZK 5,140.00Save 25%Please contact Customer Service for Availability
Quality Segment
form
solid
potency
3200 mg/kg LD50, oral (Rat)
pH
6-8 (20 °C, 10 g/L in H2O)
mp
195 °C (decomposition)
bulk density
470 kg/m3
storage temp.
no temp limit
SMILES string
[Na+].[S](=O)(=O)([O-])OCCCCCCCC
InChI
1S/C8H18O4S.Na/c1-2-3-4-5-6-7-8-12-13(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1
InChI key
WFRKJMRGXGWHBM-UHFFFAOYSA-M
Application
Matrix-assisted laser desorption/ionization: Sodium octyl sulfate was evaluated as a matrix in MALDI-TOF MS for amino acids, showcasing its potential in enhancing ionization efficiency in proteomic analyses (Yazdabadi et al., 2021).
Ultrasonic synthesis of stable oil filled microcapsules: This study explores the ultrasonic synthesis of oil-filled microcapsules using thiolated chitosan, potentially involving derivatives like ᴅʟ-N-Acetylhomocysteine thiolactone for improved stability and functional properties. The research provides insights into advanced microencapsulation techniques relevant in pharmaceutical and industrial applications (Source).
Analysis Note
Assay (acidimetric; calculated on dried substance): ≥ 99
Identity (IR-spectrum): passes test
Chloride (Cl): ≤ 0.1 %
Heavy metals (as Pb): ≤ 0.004 %
Free octanol (GC): ≤ 0.1 %
Sulfated ash: 29.0 - 32.0 %
Loss on drying (60 °C; vacuum): ≤ 1
Identity (IR-spectrum): passes test
Chloride (Cl): ≤ 0.1 %
Heavy metals (as Pb): ≤ 0.004 %
Free octanol (GC): ≤ 0.1 %
Sulfated ash: 29.0 - 32.0 %
Loss on drying (60 °C; vacuum): ≤ 1
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form powder | form crystals | form powder |
| storage temp. no temp limit | storage temp. - | storage temp. - | storage temp. - |
| mp 195 °C (decomposition) | mp 195 °C (dec.) (lit.) | mp 195 °C (dec.) (lit.) | mp - |
| Quality Level 200 | Quality Level 200 | Quality Level 100 | Quality Level 100 |
| pH 6-8 (20 °C, 10 g/L in H2O) | pH - | pH - | pH - |
| bulk density 470 kg/m3 | bulk density - | bulk density - | bulk density - |
Storage Class
11 - Combustible Solids
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


