Skip to Content
Merck

Skip To

1.12219

Lanthanum(III) chloride heptahydrate

98%, for analysis

Synonym(s):

Lanthanum(III) chloride heptahydrate 98%, Lanthanum trichloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Please contact Customer Service for Availability
CZK 3,630.00
250 g
Please contact Customer Service for Availability
CZK 5,950.00

About This Item

Empirical Formula (Hill Notation):
Cl3La · 7H2O
CAS Number:
Molecular Weight:
371.37
UNSPSC Code:
12352302
EC Index Number:
233-237-5
NACRES:
NB.21
MDL number:

CZK 3,630.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Lanthanum(III) chloride heptahydrate 98%, for analysis

Quality Segment

description

for inorganic trace analysis

assay

98%

form

solid

potency

4184 mg/kg LD50, oral (Rat)

pH

5 (25 °C, 100 g/L in H2O)

mp

91 °C (Elimination of water of crystallisation)

bulk density

900 kg/m3

storage temp.

no temp limit

SMILES string

[La](Cl)(Cl)Cl.O.O.O.O.O.O.O

InChI

1S/3ClH.La.7H2O/h3*1H;;7*1H2/q;;;+3;;;;;;;/p-3

InChI key

FDFPDGIMPRFRJP-UHFFFAOYSA-K

Analysis Note

Identity (elemental analysis): passes test
Assay (complexometric): ≥ 98 %
Ca (Calcium): ≤ 0.001 %
Mg (Magnesium): ≤ 0.0001 %
Cu (Copper): ≤ 0.0001 %
Fe (Iron): ≤ 0.0005 %
Pb (Lead): ≤ 0.0001 %
Zn (Zinc): ≤ 0.0001 %
Water-insoluble matter: ≤ 0.1 %

Other Notes

Replaces: LX0042; LX0042-3

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2035211.05326211605
assay

98%

assay

99.999% trace metals basis

assay

-

assay

99.9% trace metals basis

form

solid

form

solid

form

solid

form

powder, crystals or chunks

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

no temp limit

storage temp.

-

storage temp.

no temp limit

storage temp.

-

mp

91 °C (Elimination of water of crystallisation)

mp

91 °C (dec.) (lit.)

mp

69.9 °C

mp

92-93 °C (lit.)

description

for inorganic trace analysis

description

-

description

for inorganic trace analysis

description

-


Still not finding the right product?


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters