Skip to Content
Merck
All Photos(1)

Key Documents

1.11452

Millipore

Phthaldialdehyde

for fluorometry

Synonym(s):

Phthaldialdehyde, OPA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CZK 2,660.00
50 G
CZK 21,700.00

CZK 2,660.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
CZK 2,660.00
50 G
CZK 21,700.00

About This Item

Linear Formula:
C6H4-1,2-(CHO)2
CAS Number:
Molecular Weight:
134.13
MDL number:
UNSPSC Code:
12352114
EC Index Number:
211-402-2
NACRES:
NA.21

CZK 2,660.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

solid

autoignition temp.

480 °C

potency

178 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

7 (20 °C, 53 g/L in H2O)

bp

83-84 °C/1 hPa

mp

52-58 °C

transition temp

flash point 132 °C

solubility

53 g/L

density

1.13 g/cm3 at 20 °C

bulk density

530 kg/m3

storage temp.

2-30°C

SMILES string

O=Cc1c(cccc1)C=O

InChI

1S/C8H6O2/c9-5-7-3-1-2-4-8(7)6-10/h1-6H

InChI key

ZWLUXSQADUDCSB-UHFFFAOYSA-N

Application


  • Combined techniques for revealing the mechanism beneath the inhibition effects of pectin on gluten digestibility using static in vitro gastro-duodenal protocols.: This study uses a combination of techniques to explore how pectin affects gluten digestibility, providing insights into the interaction of phthaldialdehyde derivatives with gluten proteins (Lin et al., 2024).

  • Analysis of Gizzerosine in Foodstuffs by HPLC Involving Pre-column Derivatization with o-Phthaldialdehyde.: This research develops a method using HPLC with pre-column derivatization involving phthaldialdehyde to analyze gizzerosine in foodstuffs, demonstrating the compound′s utility in food safety studies (Li and Wu, 2024).

  • Comprehensive studies on the development of HPLC-MS/MS and HPLC-FL based methods for routine determination of homocysteine thiolactone in human urine.: This study develops and validates methods using phthaldialdehyde derivatives for the determination of homocysteine thiolactone in human urine, contributing to clinical diagnostics and biochemical research (Piechocka and Głowacki, 2024).

  • Smart-detection approach for protein residues to evaluate the cleaning efficacy of reusable medical devices.: Utilizing phthaldialdehyde for protein residue detection, this study provides a smart detection approach to improve the cleaning efficacy of medical devices, enhancing patient safety (Ouirungroj et al., 2024).

  • One-pot sample preparation procedure for the determination of protein N-linked homocysteine by HPLC-FLD based method.: This research presents a novel one-pot sample preparation method using phthaldialdehyde for determining protein N-linked homocysteine, advancing analytical techniques in protein chemistry (Piechocka and Głowacki, 2023).


Analysis Note

Purity (GC): ≥ 99 %
Identity (IR): conforms
Melting point: 54 - 58 °C
Fluorescence (340 nm, 0,8 g/l, ethanol, as quinine): ≤ 2 ppb
Phthalic acid (TLC): ≤ 0.5 %
Nitrogen compounds (as N): ≤ 0.05 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service