Skip to Content
Merck
All Photos(1)

Key Documents

1.11106

Supelco

Chloride Test Kit, titrimetric

titrimetric, for use with MCOLORTEST®

Synonym(s):

Chloride Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

200 TESTS
CZK 3,080.00

CZK 3,080.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
200 TESTS
CZK 3,080.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

CZK 3,080.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Product Name

Chloride Test, titrimetric, for use with MCOLORTEST®

specific analyte(s)

chloride

Quality Level

compatibility

for use with MCOLORTEST®

detection method

titrimetric

storage temp.

15-25°C

General description

Chloride is found in the earth´s crust and can therefore also be found in all natural waters at some concentration. This basic parameter of water analysis can be checked using our Chloride Test. This MQuant® Chloride test (formally MColortest®) is suitable for the titrimetric determination using a graduated syringe.

Method
In nitric solution chloride ions are titrated with mercury(II) nitrate solution against 1,5-diphenylcarbazone as the indicator, forming slightly dissociated mercury(II) chloride in the process. At the titration end-point, excess mercury(II) ions react with the indicator to form a blue-violet complex. The chloride concentration is determined from the consumption of titration solution (mercurimetric determination).

The test can be used in
  • Groundwater, surface water, and seawater (after dilution)
  • Waters from aquaculture
  • Drinking water and mineral water
  • Water for concrete in the construction industry
  • Industrial water and wastewater
  • Boiler and boiler feed water, cooling water
  • Swimming-pool water
  • Food after appropriate sample pretreatment
  • Soils and fertilizers after appropriate sample pretreatment
Method: titrimetric with titration pipette MQuant®

Application

  • PREFUL MRI for Monitoring Perfusion and Ventilation Changes after Elexacaftor-Tezacaftor-Ivacaftor Therapy for Cystic Fibrosis: A Feasibility Study.: This study demonstrates the use of PREFUL MRI to monitor changes in perfusion and ventilation in cystic fibrosis patients post-therapy, showcasing its potential for non-invasive assessment in clinical settings (Dohna et al., 2024).
  • Wearable sweat chloride sensors: materials, fabrication and their applications.: Reviews advances in materials and fabrication techniques for wearable sweat chloride sensors, discussing their significant potential for real-time health monitoring and diagnostics (Patel et al., 2024).
  • Initial characteristics of cystic fibrosis in Algeria: Description of 34 pediatric cases.: Provides an overview of the clinical presentation and initial characteristics of cystic fibrosis in a pediatric Algerian cohort, contributing to the regional understanding of this genetic disorder (Bendoukha et al., 2024).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1B - STOT RE 2

Target Organs

Kidney

Supplementary Hazards

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service