Skip to Content
Merck
All Photos(1)

Documents

1.10049

Supelco

Molybdenum Test Strips, colorimetric

5-250 mg/L (Mo), MQuant®

Synonym(s):

Molybdenum chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Molybdenum Test, colorimetric, 5-250 mg/L (Mo), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

molybdenum

measuring range

5-250 mg/L (Mo)

detection method

colorimetric

storage temp.

15-25°C

General description

Method: Colorimetric, with test strips and reagent, 5 - 20 - 50 - 100 - 250 mg/L (Mo), MQuant®

The main use of the MQuant® Molybdenum test is for boiler and cooling water analysis. The purpose is to determine the concentration of molybdenum or molybdate which is used as a corrosion inhibitor. The test includes test strips and all necessary additional reagents. In acidic solution molybdate ions react with toluene- 3,4-dithiol to form a green complex. The molybdenum concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the molybdenum concentration and a possible dilution factor prior to using a quantitative method such as a Spectroquant® test.

Linkage

Replaces: 10049-1; 10049

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service