Skip to Content
Merck
All Photos(1)

Key Documents

1.09994

Supelco

Vanadium standard

1000 mg V, (VOSO₄ in 8.6% H₂SO₄) Titrisol®

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
CZK 1,240.00

CZK 1,240.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 AMP
CZK 1,240.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CZK 1,240.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

Titrisol®

form

concentrate

concentration

1000 mg V in 8.6% H2SO4

technique(s)

AAS: suitable
ICP: suitable

pH

0.5 (20 °C in H2O)

density

1.06 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Features and Benefits

  • TitrisolTitrisol® offers a wide range of concentrated single-element standards.
  • Dilute the concentrated solution to achieve the desired standard concentration.
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST).
  • Supplied in easy-to-handle plastic ampoules.
  • Accompanied with a batch-specific certificate of analysis (COA) containing information on the NIST batch and minimum shelf life.
  • Provides reliable and reproducible results.
  • Analysis Note

    content (V): 1000 mg/Ampoule
    Determination method: Oxidimetric titration.
    (traceableof NIST - SRM 136e)
    Accuracy of the method: +/- 2 mg

    Legal Information

    Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

    Pictograms

    Corrosion

    Signal Word

    Warning

    Hazard Statements

    Hazard Classifications

    Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

    Storage Class Code

    8B - Non-combustible, corrosive hazardous materials

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service