Skip to Content
Merck

Skip To

1.09871

Hydrochloric acid solution

Chloride standard, 1000 mg Cl- (HCl in H2O) Titrisol®

Synonym(s):

Hydrochloric acid standard solution, Chloride standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 amp
Please contact Customer Service for Availability
CZK 1,040.00

About This Item

Linear Formula:
HCl
CAS Number:
Molecular Weight:
36.46
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
1098214
MDL number:

CZK 1,040.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Titrisol®

technique(s)

ion chromatography: suitable

pH

0.6 (20 °C in H2O)

density

1.01 g/cm3 at 20 °C

application(s)

environmental
food and beverages

format

single component solution

storage temp.

15-25°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Analysis Note

content (Cl⁻): 1000 mg / Ampoule
Determination method: Argentometric titration
(traceable to NIST - SRM 999b)
Accuracy of the method: +/- 2 mg

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.099741.099711.09973
application(s)

environmental
food and beverages

application(s)

-

application(s)

-

application(s)

-

technique(s)

ion chromatography: suitable

technique(s)

titration: suitable

technique(s)

titration: suitable

technique(s)

titration: suitable

format

single component solution

format

-

format

-

format

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

pH

0.6 (20 °C in H2O)

pH

1 ( in H2O, undiluted)

pH

<1 (20 °C in H2O)

pH

<1 (20 °C in H2O)


Still not finding the right product?

Explore all of our products under Hydrochloric acid solution


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters