Skip to Content
Merck
All Photos(1)

Key Documents

1.09635

Supelco

Ethylbenzene

reference substance for gas chromatography

Synonym(s):

Ethylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
CZK 6,040.00

CZK 6,040.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
CZK 6,040.00

About This Item

Empirical Formula (Hill Notation):
C8H10
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-849-4
NACRES:
NA.24

CZK 6,040.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

9.5 hPa ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

430 °C

potency

3500 mg/kg LD50, oral (Rat)
15354 mg/kg LD50, skin (Rabbit)

expl. lim.

1.0-7.8 % (v/v)

technique(s)

gas chromatography (GC): suitable

kinematic viscosity

0.63 cSt(40 °C)

bp

136 °C/1013 hPa

mp

-95 °C

transition temp

flash point 15 °C

solubility

0.2 g/L

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(C)c1ccccc1

InChI

1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

YNQLUTRBYVCPMQ-UHFFFAOYSA-N

Application


  • Ethylbenzene in Environmental Biodegradation: Research on the genomic diversity of Pseudomonas species capable of degrading hydrocarbons like ethylbenzene illustrates its role in bioremediation, showcasing how these bacteria can be utilized to address environmental contamination (Imperato et al., 2019).

  • Toxicokinetic Modeling of Ethylbenzene: A study focused on the impact of biological and environmental variabilities on biological monitoring highlights the importance of ethylbenzene in toxicokinetic models, enhancing our understanding of its behavior in occupational health scenarios (Berthet et al., 2010).

  • Health Impact Studies: Examination of relationships between environmental exposures to volatile organic compounds, including ethylbenzene, and biochemical liver tests applies canonical correlation analysis, aiding in the assessment of potential health risks (Liu et al., 2009).

  • Advanced Sensor Technology: The development of fully integrated sensor systems that utilize ethylbenzene as a test analyte in liquid-phase chemical detection, demonstrating the compound′s utility in enhancing sensor technology for analytical chemistry applications (Vancura et al., 2007).

Analysis Note

Purity (GC): ≥ 99.5
Identity (IR-spectrum): conforms

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - STOT RE 2

Target Organs

hearing organs

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service