Skip to Content
Merck
All Photos(1)

Documents

1.08641

Supelco

Water detection paste N

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401
NACRES:
NB.21

pH

11 ( in H2O, suspension)

Quality Level

density

1.6 g/cm3 at 20 °C

storage temp.

no temp limit

Analysis Note

Suitability for water detection: conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service