Skip to Content
Merck

Skip To

1.08374

L-Tryptophan

for biochemistry

Synonym(s):

L-Tryptophan, Trp

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Please contact Customer Service for Availability
CZK 1,400.00
100 g
Please contact Customer Service for Availability
CZK 5,060.00
500 g
Please contact Customer Service for Availability
CZK 20,700.00
10 kg
Please contact Customer Service for Availability
CZK 196,700.00

About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
UNSPSC Code:
12352209
EC Index Number:
200-795-6
NACRES:
NA.21
MDL number:

CZK 1,400.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

potency

16000 mg/kg LD50, oral (Rat)

pH

5.5-7.0 (20 °C, 10 g/L in H2O)

mp

290 °C (decomposition)

solubility

10 g/L

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

[nH]1c2c(c(c1)CC(N)C(=O)O)cccc2

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)

InChI key

QIVBCDIJIAJPQS-UHFFFAOYSA-N

Application


  • Biotechnological production of L-Tryptophan: The study by Nefedova et al. explores the molecular mechanisms in metal oxide nanoparticle interactions with L-Tryptophan, highlighting its significance in nanoparticle synthesis, which is crucial for biotechnological applications in pharmaceutical manufacturing and life sciences (Nefedova et al., 2024).

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): 99.0 - 101.0 %
Identity (IR-spectrum): passes test
Identity (specific rotation): passes test
Spec. rotation (α 20/D, 10 g/l, water, calc. on dried substance): -30.0 to -32.5
Appearance: white to almost white, crystalline or amorphous powder
Appearance of solution (10 g/l, Hydrochloric acid 1 mol/l): clear and not more intense in color than reference solution BY6
Chloride (Cl): ≤ 200 ppm
Sulfate (SO₄): ≤ 200 ppm
Fe (Iron): ≤ 20 ppm
Ninhydrin-positive substances (LC)(any ninhydrin-positive impurity): ≤ 0.2 %
Ninhydrin-positive substances (LC) (ammonium (570 nm)): ≤ 0.02 %
Ninhydrin-positive substances (LC)(total impurities): ≤ 0.5 %
Loss on drying (105 °C; 3 h): ≤ 0.2 %

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.08396PHR1176T2610000
form

solid

form

solid

form

-

form

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-8°C

mp

290 °C (decomposition)

mp

290 °C (decomposition)

mp

-

mp

280-285 °C (dec.) (lit.)

Quality Level

100

Quality Level

500

Quality Level

300

Quality Level

-

solubility

10 g/L

solubility

10 g/L

solubility

-

solubility

-

potency

16000 mg/kg LD50, oral (Rat)

potency

16000 mg/kg LD50, oral (Rat)

potency

-

potency

-


Still not finding the right product?

Explore all of our products under L-Tryptophan


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters