Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
500 g | Please contact Customer Service for Availability | CZK 3,340.00 |
About This Item
form
medium granules (dehydrated (DCM))
Quality Segment
manufacturer/tradename
GranuCult®
technique(s)
microbiological culture: suitable
application(s)
food and beverages, microbiology
storage temp.
15-25°C
suitability
Escherichia coli, coliforms
General description
Analysis Note
Appearance (colour): green
pH-value (25 °C): 7.0 - 7.4
Recovery on test medium (Escherichia coli ATCC 25922 (WDCM 00013)): ≥ 70 %
Recovery on test medium (Citrobacter freundii ATCC 8090): ≥ 70 %
Recovery on test medium (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): ≥ 70 %
Medium colour (Escherichia coli ATCC 25922 (WDCM 00013)): yellow
Medium colour (Citrobacter freundii ATCC 8090): yellow
Medium colour (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): blue
Colony colour (Escherichia coli ATCC 25922 (WDCM 00013)): yellow-orange
Colony colour (Citrobacter freundii ATCC 8090): yellow-orange
Colony colour (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): red
Growth (Bacillus cereus ATCC 11778 (WDCM 00001)): inhibited
Incubation: 24 hrs.; 35 °C; aerobic.
Other Notes
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) microbiological culture: suitable | technique(s) microbe id | utilization test: suitable, microbiological culture: suitable | technique(s) microbiological culture: suitable | technique(s) microbiological culture: suitable |
| suitability Escherichia coli, coliforms | suitability Enterobacter spp., Escherichia coli, Proteus spp., Salmonella spp., Staphylococcus spp., coliforms, lactose-fermenting bacteria | suitability aerobic bacteria (total count) | suitability anaerobic bacteria |
| form medium granules (dehydrated (DCM)) | form medium granules (dehydrated (DCM)) | form medium granules (dehydrated (DCM)) | form granular ( ) |
| application(s) food and beverages, microbiology | application(s) food and beverages, microbiology | application(s) food and beverages, microbiology | application(s) food and beverages |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Still not finding the right product?
Explore all of our products under Lactose TTC Agar with Tergitol™ 7
Storage Class
11 - Combustible Solids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


