Skip to Content
Merck

Skip To

1.07563

Extran® AP 12

greener alternative

powder, alkaline

Synonym(s):

Extran® AP 12 cleaning agent; Extran® AP 12 detergent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2 kg
Please contact Customer Service for Availability
CZK 3,310.00
10 kg
Please contact Customer Service for Availability
CZK 11,900.00
25 kg
Please contact Customer Service for Availability
CZK 23,800.00

About This Item

NACRES:
NA.21
UNSPSC Code:
41103206

CZK 3,310.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

pH

12.3 (20 °C, 3 g/L in H2O)

bulk density

900 kg/m3

greener alternative category

storage temp.

no temp limit

General description

This product is identical to EX0990 with the same quality and specification. Some pack sizes have changed to provide a globally harmonized offering.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is environmentally friendly having bio-degradable active ingredients. Click here for more information.

Analysis Note

Identity: passes test
pH-value (1 %; water): ≥ 12.5
Foaming volume (5 %; water): ≤ 1 ml
Bulk density: about 90

Other Notes

Replaces: EX0990-1

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.075701.075591.07558
form

solid

form

-

form

liquid

form

solid

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

no temp limit

Quality Level

300

Quality Level

300

Quality Level

200

Quality Level

300

pH

12.3 (20 °C, 3 g/L in H2O)

pH

11.4 (20 °C, 3 g/L in H2O)

pH

2.0 (20 °C, 2 g/L in H2O)

pH

11.3 (20 °C, 3 g/L in H2O)

bulk density

900 kg/m3

bulk density

750 kg/m3

bulk density

-

bulk density

700 kg/m3

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product


pictograms

Corrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters