Skip to Content
Merck

Skip To

1.06733

Sodalime

with indicator, granules ~ 1 - 2.5 mm

Synonym(s):

Sodalime granules; Calcium sodium carbonate; Soda-lime

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 g
Please contact Customer Service for Availability
CZK 2,410.00
2.5 kg
Please contact Customer Service for Availability
CZK 7,680.00

About This Item

NACRES:
NB.51
UNSPSC Code:
12352402

CZK 2,410.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

granular

matrix

calcium hydroxide base material

particle size

1-2.5 mm

bulk density

750 kg/m3

storage temp.

no temp limit

Application

Sodalime is a mixture of NaOH and CaO. It is used as an excellent drying agent or desiccant in several industries. It is also used as a CO2 absorber.

Analysis Note

Identity: passes test
Absorption capacity for CO₂ (according to NF): ≥ 25 %
Bulk density: about 85

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
26643472073432504
particle size

1-2.5 mm

particle size

+100 mesh

particle size

2-5 mm

particle size

-

form

granular

form

granular

form

pellets

form

solid

matrix

calcium hydroxide base material

matrix

-

matrix

-

matrix

-

bulk density

750 kg/m3

bulk density

-

bulk density

~75 g/100 mL

bulk density

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

storage temp.

no temp limit

storage temp.

-

storage temp.

-

storage temp.

-


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters