Skip to Content
Merck

Skip To

1.06529

Sodium polyphosphate

(Graham′s salt) EMPLURA®

Synonym(s):

Sodium polyphosphate, Graham′s salt, Sodium metaphosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kg
Please contact Customer Service for Availability
CZK 1,380.00
5 kg
Please contact Customer Service for Availability
CZK 4,720.00
50 kg
Please contact Customer Service for Availability
CZK 35,200.00

About This Item

Linear Formula:
(NaPO3)n
CAS Number:
UNSPSC Code:
12352302
EC Index Number:
233-782-9
NACRES:
NA.21
MDL number:
Form:
solid
Solubility:
1000 g/L

CZK 1,380.00


Please contact Customer Service for Availability
Packaging: HDPE bottle

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

EMPLURA®

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

concentration

≥68% ( P2O5, acidimetric)

pH

5.7 (20 °C, 10 g/L in H2O)

mp

550 °C

solubility

1000 g/L

density

2.5 g/cm3 at 20 °C

bulk density

1000 kg/m3

anion traces

chloride (Cl-): ≤0.02%

cation traces

Fe: ≤0.01%, heavy metals (as Pb): ≤0.005%

storage temp.

2-30°C

SMILES string

[Na+].[P](=O)(=O)[O-]

InChI

1S/Na.HO3P/c;1-4(2)3/h;(H,1,2,3)/q+1;/p-1

InChI key

AQMNWCRSESPIJM-UHFFFAOYSA-M

Application


  • Novel method for the preparation of controlled-release theophylline granules coated with a polyelectrolyte complex of sodium polyphosphate-chitosan.: This paper presents a novel approach to creating controlled-release theophylline granules using a coating made from a polyelectrolyte complex of sodium polyphosphate and chitosan. This method could significantly impact pharmaceutical formulations by improving drug release profiles and patient compliance (Kawashima et al., 1985).

Analysis Note

Assay (acidimetric, P₂O₅): ≥ 68 %
Identity: passes test
Chloride (Cl): ≤ 0.02 %
Heavy metals (as Pb): ≤ 0.005 %
Fe (Iron): ≤ 0.01 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.0660971600305553
form

solid

form

solid

form

solid

form

crystalline

solubility

1000 g/L

solubility

545 g/L

solubility

-

solubility

-

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

anion traces

chloride (Cl-): ≤0.02%

anion traces

chloride (Cl-): ≤0.01%

anion traces

-

anion traces

-

density

2.5 g/cm3 at 20 °C

density

1.2 g/cm3 at 20 °C

density

-

density

-

pH

5.7 (20 °C, 10 g/L in H2O)

pH

3.5-3.8 (20 °C, 100 g/L in H2O)

pH

-

pH

-


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Bonjour, Connaissez-vous le nombre d'unité moyen (n) du Sodium Polyphosphate ? Merci, Cordialement

    1 answer
    1. This product is commonly referred to as Graham′s salt. The literature suggests that typically the n is 6, to make it sodium hexametaphosphate. However, this is not included in the product testing nor has this been verified. The end user would have to verify this empirically.

      Helpful?

  4. Is it polydisperse or mono disperse?

    1 answer
    1. This product is Graham's salt, in which typically n=6, to make it sodium hexametaphosphate, and in the form of a cyclic hexamer. This should make it a relatively monodisperse product. However, this is not included in the testing specifications, so it cannot be confirmed. This would need to be verified empirically.

      Helpful?

Reviews

No rating value

Active Filters