Skip to Content
Merck
All Photos(2)

Key Documents

1.05983

Supelco

D-Mannitol

for the determination of boric acid

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
CZK 1,750.00
1 KG
CZK 5,100.00

CZK 1,750.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
CZK 1,750.00
1 KG
CZK 5,100.00

About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-711-8
NACRES:
SE.11

CZK 1,750.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

autoignition temp.

410 °C

potency

13500 mg/kg LD50, oral (Rat)

pH

5-7 (20 °C, 100 g/L in H2O)

bp

290-295 °C/4 hPa

mp

167-170 °C (lit.)

solubility

213 g/L

bulk density

400‑500 kg/m3

storage temp.

no temp limit

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • D-mannitol sensor based on molecularly imprinted polymer on electrode modified with reduced graphene oxide decorated with gold nanoparticles.: This study presents a mannitol sensor utilizing a molecularly imprinted polymer on a substrate modified with reduced graphene oxide and gold nanoparticles, emphasizing advances in analytical chemistry for precise detection of mannitol (Stradiotto et al., 2017).

  • Borate-polyol complexes in aqueous solution: determination of enthalpies by thermometric titrimetry.: Investigating the interactions between borate and polyols like mannitol, this research applies thermometric titrimetry to assess complex formation, providing key thermodynamic data relevant for analytical chemistry (Aruga, 1985).

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Analysis Note

Identity (IR-spectrum): passes test
Melting point: 166 - 169
Suitability for boric acid determination: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service