1.05934
Manganese(II) chloride dihydrate
for analysis EMSURE®
Synonym(s):
Manganese(II) chloride dihydrate, Manganese dichloride
About This Item
Quality Level
product line
EMSURE®
Assay
≥99.0% (complexometric)
form
solid
potency
250 mg/kg LD50, oral (Rat)
pH
5-6 (20 °C, 50 g/L in H2O)
bp
1190 °C (anhydrous substance)
mp
650 °C (anhydrous substance)
solubility
1200 g/L
density
2.27 g/cm3 at 20 °C
bulk density
850 kg/m3
anion traces
sulfate (SO42-): ≤0.002%
cation traces
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.001%
Mg: ≤0.005%
Na: ≤0.005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Zn: ≤0.001%
storage temp.
2-30°C
InChI
1S/2ClH.Mn.2H2O/h2*1H;;2*1H2/q;;+2;;/p-2
InChI key
BEYCFZBNRLPHEP-UHFFFAOYSA-L
Analysis Note
Sulfate (SO₄): ≤ 0.002 %
Ca (Calcium): ≤ 0.001 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.001 %
Mg (Magnesium): ≤ 0.005 %
Na (Sodium): ≤ 0.005 %
Ni (Nickel): ≤ 0.0005 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.001 %
Substances reducing potassium permanganate: ≤ 0.0005 %
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2
Target Organs
Brain
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point(C)
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service