Skip to Content
Merck
All Photos(1)

Documents

1.05705

Supelco

Molecular sieve 0.5 nm

beads ~ 2 mm

Synonym(s):

Molecular sieve 0.5 nm, Sodium aluminium silicate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41123003
EC Index Number:
215-283-8

Quality Level

form

beads

surface area

800 m2/g

loss

≤7.5% loss on ignition, 950°C

particle size

~2 mm

pore size

0.5 nm

pH

8-11 ( in H2O)

mp

>1600 °C

bulk density

700‑750 kg/m3

storage temp.

no temp limit

Analysis Note

Identity: passes test
Loss on ignition (950 °C): ≤ 7.5 %
Water absorption capacity (24 hrs.,80 % relative humidity): ≥ 20 %

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service