Skip to Content
Merck
All Photos(1)

Documents

1.05226

Sigma-Aldrich

New fuchsin (C.I. 42520)

for microscopy Certistain®

Synonym(s):

New fuchsin (C.I. 42520)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24ClN3
CAS Number:
Molecular Weight:
365.90
MDL number:
UNSPSC Code:
41116130
EC Index Number:
221-831-7

Quality Level

form

solid

composition

Dye, ≥75% spectrophotometric assay

pH

3-4 (20 °C, 10 g/L in H2O)

solubility

10 g/L

bulk density

450 kg/m3

εmax

1810-2430 at 553-556 nm in 50% ethanol at 0.003 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C22H24N3.ClH/c1-13-10-16(4-7-19(13)23)22(17-5-8-20(24)14(2)11-17)18-6-9-21(25)15(3)12-18;/h4-12H,23-25H2,1-3H3;1H/q+1;/p-1

InChI key

ORDGVBRMUJCHEO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

The dry dye New fuchsin (C.I. 42520) for microscopy Certistain®, is used for the preparation of a staining solution that can be used for staining of histological tissue sections or other samples of human origin according to Ziehl-Neelsen, Gram staining as well as in combination with Schiff′s reagent. As a Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. New fuchsin belongs to the group of triaminotriphenylmethane dyes.
The dye is used in Ziehl-Neelsen and Gram staining as well as for methods with Schiff′s reagent.
The cell wall of acid-fast bacteria (AFB) has a high proportion of wax and lipids and hence absorbs dyes only very slowly. As a measure to enhance the absorption of the fuchsin dye and thus the formation of the mycolate-fuchsin complex in the cell wall, the carbolfuchsin solution applied to the specimen is normally heated to evaporation point. If the Kinyoun carbolfuchsin solution is used, the heating of the solution is dispensable. Consequently, the release of hazardous phenolic vapors will be avoided.
Once the AFB have absorbed the fuchsin dye, it is virtually impossible to decolorize them again, even when they are intensively treated with a decolorizing solution such as e. g. hydrochloric acid in ethanol. Accordingly, AFB are termed as acid- and alcohol-fast for staining, and are stained red in the microscopic visualization. Correspondingly, all non-acid-fast microorganisms are counterstained with an appropriate dye.
Pretreatment of the specimens with Sputofluol® dissolves the bacteria from the surrounding viscid sputum and cell material. Sputofluol also has a disinfectant effect, with the result that any microorganisms that are present are killed off. It is registered as IVD and certified as CE product. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (spectrophotometrically): ≥ 75 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (ethanol 50 %): 553 - 556 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; ethanol 50 %): 1810 - 2430
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany
SPUTOFLUOL is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service