Skip to Content
Merck

Skip To

1.04394

n-Hexane

Sign In to View Organizational & Contract Pricing.



vapor pressure

160 hPa ( 20 °C)

Quality Segment

product line

Prepsolv

assay

≥95.0% (GC)

form

liquid

autoignition temp.

240 °C

potency

25000 mg/kg LD50, oral (rat), >2000 mg/kg LD50, skin (rabbit)

expl. lim.

1.0-8.1 % (v/v)

impurities

≤0.0002 meq/g Acidity, ≤0.0002 meq/g Alkalinity, ≤0.01% Water

evapn. residue

≤5.0 mg/L

transmittance

220 nm, ≥50%, 250 nm, ≥98%

kinematic viscosity

0.50 cSt(20 °C)

bp

69 °C/1013 hPa

mp

-94.3 °C

transition temp

flash point -22 °C

solubility

0.0095 g/L

density

0.66 g/cm3 at 20 °C

General description

PrepSolv solvents are tailored to the requirements of preparative HPLC to facilitate scale-up from analytical to preparative separations. With their extremely low evaporation residue (< 1 mg/l) and low water content, they ensure optimal protection for columns.

Application

for preparative chromatography Prepsolv

Preparation Note

Store at +5°C to +30°C.

Analysis Note

Purity (GC):≥ 95.0 %
Identity (IR):conforms
Evaporation residue:≤ 5.0 mg/l
Water:≤ 0.01 %
Acidity:≤ 0.0002 meq/g
Alkalinity:≤ 0.0002 meq/g
Transmission (at 220 nm):≥ 50 %
Transmission (from 250 nm):≥ 98 %

Legal Information

PREPSOLV is a trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

flash_point_f

-7.6 °F - closed cup

flash_point_c

-22 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

110-54-3

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters