Skip to Content
Merck

1.04238

Ponceau S (C.I. 27195)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H12N4Na4O13S4
CAS Number:
Molecular Weight:
760.57
NACRES:
NA.21
MDL number:
UNSPSC Code:
12171500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Quality Segment

composition

Dye, ≥80% spectrophotometric assay

pH

9.0 (20 °C, 10 g/L in H2O)

solubility

10 g/L

bulk density

660 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C22H16N4O13S4.4Na/c27-22-20(43(37,38)39)10-12-9-16(41(31,32)33)6-7-17(12)21(22)26-25-18-11-14(3-8-19(18)42(34,35)36)24-23-13-1-4-15(5-2-13)40(28,29)30;;;;/h1-11,25H,(H,28,29,30)(H,31,32,33)(H,34,35,36)(H,37,38,39);;;;/q;4*+1/p-4/b24-23?,26-21-;;;;

InChI key

LZYGEJPERQQCIY-STNZDNLRSA-J

General description

The dry dye Ponceau S (C.I. 27195) for microscopy Certistain®, is used for the preparation of a staining solution for staining connective tissue in histological sections of human origin. As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.

Application

for microscopy Certistain®

Analysis Note

Dye content (spectrophotometrically): ≥ 80 %
Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 517 - 523 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; water): 460 - 600
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Ponceau S (C.I. 27195)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library