Skip to Content
Merck
All Photos(1)

Key Documents

1.04187

Sigma-Aldrich

Amido black 10 B (C.I.20470)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14N6Na2O9S2
CAS Number:
Molecular Weight:
616.49
MDL number:
NACRES:
NA.41

form

solid

Quality Level

potency

>5000 mg/kg LD50, oral (Rat)

composition

Dye, ≥80% spectrophotometric assay

pH

9 (25 °C, 10 g/L in H2O)

solubility

10 g/L

bulk density

670 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C22H16N6O9S2.2Na/c23-19-18-12(11-17(39(35,36)37)21(22(18)29)27-24-13-4-2-1-3-5-13)10-16(38(32,33)34)20(19)26-25-14-6-8-15(9-7-14)28(30)31;;/h1-11,24H,23H2,(H,32,33,34)(H,35,36,37);;/q;2*+1/p-2/b26-25?,27-21+;;

InChI key

HKBVRFLHNUEVRO-DWTBGCDMSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Amido black 10 B (C.I.20470) for electrophoresis, is used for staining connective tissue, in electrophoresis, and as a redox indicator. Proteins possess free amino and carboxyl groups and are charged positively or negatively in aqueous solutions, depending on the pH. In an alkaline milieu they are negatively charged and migrate in the electric field towards the anode at a speed that depends on the number of charges they carry. This difference in migration velocities makes it possible to separate the serum protein mixture into groups of individual components. The separated protein fractions are stained with amido black and measured photometrically or densitometrically. As a rule, five fractions are observed after staining - albumin, alpha 1- and alpha 2-globulins, beta 1- and beta 2-globulins, and beta-globulins - where albumin has the highest migration velocity and gamma-globulin the lowest. Differentiation of serum proteins by electrophoresis is important for the diagnosis and prognosis of numerous diseases. The dye has a constant dye content and a reliable high quality, which is one of the major precondition for a reproducible diagnostic staining.

Application

for electrophoresis

Analysis Note

Dye content (spectrophotometrically): ≥ 80 %
Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 617 - 620 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; water): 750 - 960
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for electrophoresis: passes test
Suitability as redox indicator: passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service