Skip to Content
Merck
All Photos(1)

Documents

1.04186

Sigma-Aldrich

Nitro blue tetrazolium chloride (NBT)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
817.64
MDL number:
UNSPSC Code:
41116130
NACRES:
NA.21

form

solid

mp

189 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C40H30N10O6.2ClH/c1-55-37-25-29(13-23-35(37)47-43-39(27-9-5-3-6-10-27)41-45(47)31-15-19-33(20-16-31)49(51)52)30-14-24-36(38(26-30)56-2)48-44-40(28-11-7-4-8-12-28)42-46(48)32-17-21-34(22-18-32)50(53)54;;/h3-26H,1-2H3;2*1H/q+2;;/p-2

InChI key

FSVCQIDHPKZJSO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nitro blue tetrazolium chloride (NBT) is a dry dye for the staining of histological tissue sections and histochemical as well as in vivo methods on material of human origin. The dye has a constant dye content and a reliable high quality, which is one of the major precondition for a reproducible diagnostic staining.

Application

for microscopy

Analysis Note

Identity (UV/VIS-Spectrum)passes testAbsorption maximum λ max (Methanol)256 - 259 nmSpec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; methanol; calc. on dried substance)800 - 900Loss on drying (Vacuum < 7 mbar, P₂O₅, 24 h)≤ 12 %Suitability for microscopypasses test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service