Skip to Content
Merck
All Photos(1)

Key Documents

1.03866

Millipore

GranuCult′ Prime Tryptose Broth

acc. t FDA-BAM

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
CZK 4,780.00

CZK 4,780.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
500 G
CZK 4,780.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

CZK 4,780.00


Please contact Customer Service for Availability

Request more information

Quality Level

form

powder

technique(s)

microbiological culture: suitable

application(s)

food and beverages
pharmaceutical

suitability

nonselective for

General description

GranuCult® prime Tryptose Broth is a nutrient rich broth medium designed for the cultivation of fastidious microorganisms from food and animal feed, water and other samples. This culture medium contains tryptose which is a mixture of tryptic digested proteins. It provides nitrogen, vitamins and amino acids. Glucose acts as a carbon source whilst sodium chloride maintains the osmotic balance.

Upon completion of incubation, the inoculum can be streaked onto specific agar plates for isolation and detection.

Application

GranuCult® prime Tryptose Broth is used to enrich and cultivate pathogenic microorganisms like streptococci, pneumococci etc. in test samples.
According to FDA-BAM Chapter No. 10 Listeria, Tryptose broth is used for subculturing before serological subtyping of Listeria monocytogenes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service