Skip to Content
Merck
All Photos(1)

Key Documents

1.03864

Millipore

GranuCult® plus Standard I Nutrient Agar

for culturing of fastidious microorganisms

Synonym(s):

NA, Granulated Nutrient Agar

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
CZK 4,500.00

CZK 4,500.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
500 G
CZK 4,500.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

CZK 4,500.00


Please contact Customer Service for Availability

Request more information

Quality Level

form

powder

storage condition

dry media

technique(s)

microbiological culture: suitable

pH

7.5±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

15-25°C

suitability

nonselective for

General description

GranuCult® plus Standard I Nutrient Agar is a granulated culture medium for cultivation of fastidious bacteria. It can also serve as high-grade base for the preparation of special culture media by adding blood, ascites fluid, serum or other enrichment. This culture medium contains peptone and yeast extract for providing nitrogen, vitamins, amino acids and carbon sources. Glucose is an additional carbon source. Sodium chloride maintains the osmotic balance and agar is the solidifying agent.

Application

Standard I Nutrient Agar can be used for the cultivation of fastidious microorganisms from food and animal feed, water and other samples. This culture medium can also be used for enumeration, isolation and enrichment of bacteria.

Features and Benefits

  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment
  • Excellent wettability, solubility, and free flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions
  • High batch-to- batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allowing many supplements to be included, with no need to add these separately

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service