Skip to Content
Merck

Skip To

1.03587

Dimethyl sulfoxide-d6

with TMS (0.1 vol.%), deuteration degree min. 99.9% for NMR spectroscopy MagniSolv

Synonym(s):

DMSO deuterated, (Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Please contact Customer Service for Availability
CZK 14,600.00

About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
UNSPSC Code:
12142201
NACRES:
NA.22
EC Index Number:
218-617-0
MDL number:
Beilstein/REAXYS Number:
1237248
Mass shift:
M+6
Form:
liquid

CZK 14,600.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.42 mmHg ( 20 °C)

Quality Segment

form

liquid

autoignition temp.

573 °F

potency

14500 mg/kg LD50, oral (Rat), 40000 mg/kg LD50, skin (Rat)

expl. lim.

42 %

technique(s)

NMR: suitable

impurities

≤0.02% Water (H<SUB>2</SUB>O) (NMR), ≤0.03% Water (H<SUB>2</SUB>O + D<SUB>2</SUB>) (Karl Fischer)

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C

transition temp

flash point 88 °C

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

storage temp.

2-30°C

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Features and Benefits

Saturation Concentration in Air: 8.0 g/m3 (20 °C)

Analysis Note

Deuteration degree: ≥ 99.9 %
Water
- K. Fischer, H₂O + D₂O:
≤ 0.03 %
Tetramethylsilane (NMR-spectrum): 0.1
Water
- NMR, H₂O:
≤ 0.02 %
Performance test (NMR-spectrum): passes test

Other Notes

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.03424296147417939
mass shift

M+6

mass shift

M+6

mass shift

M+6

mass shift

M+6

technique(s)

NMR: suitable

technique(s)

NMR: suitable

technique(s)

NMR: suitable

technique(s)

NMR: suitable

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

100

form

liquid

form

liquid

form

liquid

form

liquid

bp

189 °C (lit.)

bp

189 °C (lit.)

bp

189 °C (lit.)

bp

189 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)


Still not finding the right product?

Explore all of our products under Dimethyl sulfoxide-d6


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

190.4 °F

flash_point_c

88 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters