Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
description
shipped with cooling packs
product line
Cellvento®
form
dry powder
technique(s)
cell culture | mammalian: suitable (suspension)
application(s)
pharma/biopharma processes
storage temp.
2-8°C
General description
Application
- Specifically developed to support optimal cell growth and performance of DHFR-negative CHO suspension cell types, primarily recombinant CHO-DG44 cell lines. May be suitable for use with other CHO cell lines.
- Intended as a companion feed supplement with the production medium Cellvento® CHO-210 in fed-batch manufacturing processes.
- For research use or manufacturing purposes (not for use in humans or therapy).
Features and Benefits
- Contains only non-animal origin components, supporting regulatory compliance and reducing variability.
- Supports high cell growth and productivity.
- Easy to store and prepare
Preparation Note
Legal Information
Storage Class
11 - Combustible Solids
WGK
WGK 2
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
7790-84-3
Stay compliant with GMP using the Emprove® Program
Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.
Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.
Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite
Not finding what you are looking for? Visit the Emprove® Program Help page.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Difficulty Finding Your Product Or Lot/Batch Number?
How to Find the Product Number
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.