Skip to Content
Merck

Skip To

1.02376

Cellvento® BHK-200

serum-free, dry powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
216.658 g

Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 2,140.00
100 L

Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 17,100.00

About This Item

UNSPSC Code:
41106213

CZK 2,140.00


Estimated to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

shipped with cooling packs

product line

Cellvento®

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

application(s)

pharma/biopharma processes

shipped in

ambient

storage temp.

2-8°C

General description

Cellvento® BHK-200 medium is a serum-free cell culture medium, formulated without any animal derived component and optimized for the culture of suspension BHK21 cells at high-density and viability and efficient propagation of viruses. BHK21 cells can be grown as suspension cultures in T-flasks, shaker flasks, spinner bottles or stirred tank bioreactors.

Application

  • Cellvento® BHK-200 medium has been developed for BHK-21 cell line but is not limited to this application and may suit other vaccine producing cell lines as well.
  • Qualified for instance for the production of Foot and Mouth Disease virus (FMDV).
  • When the density of BHK21 cells satisfies process requirement, the virus can be inoculated directly to the reactor without media exchange or cell separation/ sedimentation steps, resulting in highly efficient virus production.
  • It is recommended to use only 70% of the maximum bioreactor volume for cell growth. After cells have reached the desired cell density, 30% fresh medium can be added in the reactor before inoculation and propagation of the virus.

Features and Benefits

  • Serum-free, animal-component-free formulation for improved process consistency.
  • Chemically defined components to enable better batch-to-batch reproducibility and easier regulatory documentation.
  • Optimized for compatibility with standard BHK cell lines and common feeds/additives used in upstream processing.

Preparation Note

Formulated to contain 21.6658 grams of powder per liter of medium.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.251951.2519314390C
technique(s)

cell culture | mammalian: suitable (suspension)

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable (suspension)

technique(s)

cell culture | mammalian: suitable (suspension)

form

dry powder

form

dry powder

form

liquid

form

liquid

product line

EX-CELL®

product line

Cellvento®

product line

Cellvento®

product line

Cellvento®

shipped in

ambient

shipped in

-

shipped in

-

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

-


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters