This product is powdered wood charcoal and is not activated.
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
25 kg | Please contact Customer Service for Availability | CZK 11,500.00 |
About This Item
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
11101522
EC Index Number:
231-153-3
NACRES:
NB.51
Quality Segment
form
powder
ign. residue
≤7% (600 °C)
particle size
<0.15 mm (~99%)
mp
3550 °C
bulk density
410 kg/m3
storage temp.
no temp limit
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Analysis Note
Residue on ignition (600 °C): ≤ 7 %
Particle size (< 0.15 mm): about 99%
Carbonization: conforms
Particle size (< 0.15 mm): about 99%
Carbonization: conforms
1 of 1
This Item | |||
|---|---|---|---|
| particle size <0.15 mm (~99%) | particle size - | particle size - | particle size -100 (mesh) |
| form powder | form powder | form powder | form powder |
| bulk density 410 kg/m3 | bulk density - | bulk density 150‑440 kg/m3 | bulk density - |
| mp 3550 °C | mp 3550 °C (lit.) | mp 3550 °C | mp 3550 °C (lit.) |
| Quality Level 200 | Quality Level 100 | Quality Level 200 | Quality Level 100 |
| storage temp. no temp limit | storage temp. - | storage temp. no temp limit | storage temp. - |
Storage Class
4.2 - Pyrophoric and self-heating hazardous materials
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is product number 1022049025 activated wood charcoal?
1 answer-
Helpful?
-


