Skip to Content
Merck

Skip To

1.01867

Calcium chloride dihydrate

crystalline, Ph. Eur., JP, USP, FCC, E 509

Synonym(s):

Calcium dichloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kg

Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 1,680.00
3 x 1 kg

Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 5,700.00

About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Index Number:
233-140-8
E Number:
E509
UNSPSC Code:
12352302
MDL number:

CZK 1,680.00


Estimated to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

JP, Ph. Eur., USP

reg. compliance

FCC

form

crystalline, solid

pH

4.5-8.5 (20 °C, 50 g/L in H2O)

mp

176 °C (dec.) (lit.)

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Calcium chloride dihydrate is a mineral salt typically used as buffering agent in pharmaceutical formulations. Beyond, calcium chloride dihydrate is used as a complexing builder, flavor enhancer and stabilizer for food and beverage.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.420021.371011.72580
application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical

application(s)

pharma/biopharma processes

agency

JP, USP, Ph. Eur.

agency

BP, ChP, JP, Ph. Eur., USP

agency

BP, JP, Ph. Eur., USP

agency

-

form

crystalline, solid

form

solid

form

solid

form

solid

mp

176 °C (dec.) (lit.)

mp

176 °C (dec.) (lit.)

mp

176 °C (dec.) (lit.)

mp

176 °C (dec.) (lit.)

pH

4.5-8.5 (20 °C, 50 g/L in H2O)

pH

4.5-8.5 (20 °C, 50 g/L in H2O)

pH

4.5-8.5 (20 °C, 50 g/L in H2O)

pH

4.5-8.5 (20 °C, 50 g/L in H2O)

reg. compliance

FCC

reg. compliance

FCC

reg. compliance

-

reg. compliance

FCC


Still not finding the right product?

Explore all of our products under Calcium chloride dihydrate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters