Skip to Content
Merck
All Photos(1)

Documents

1.01747

Supelco

Arsenic Test, photometric

photometric, 0.001-0.100 mg/L (As), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Arsenic Test, photometric, 0.001-0.100 mg/L (As), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 30 tests

specific analyte(s)

arsenic

measuring range

0.001-0.100 mg/L (As)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Silver DDTC)

storage temp.

15-25°C

General description

Method: photometric 0.001 - 0.100 mg/l As Spectroquant®


The method is analogous to EPA 206.4, APHA 3500-As B, and ASTM D2972-15 A.


This Spectroquant® Arsenic Reagent Test allows the accurate quantification of the arsenic content in aqueous samples.


Method applied: In acidic solution arsenic compounds are reduced by a reducing-agent mixture consisting of zinc, tin(II) chloride, and potassium iodide to form arsine which reacts in an absorption tube with silver diethyldithiocarbamate to form a red compound that is determined photometrically.


The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.


All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT RE 1 Oral - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system, Thyroid

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service