Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
50 g | Please contact Customer Service for Availability | CZK 5,010.00 |
500 g | Please contact Customer Service for Availability | CZK 25,600.00 |
About This Item
Quality Segment
description
for inorganic trace analysis
assay
99.995%
form
solid
potency
118 mg/kg LD50, oral (Rat)
pH
5-8 (20 °C, 50 g/L in H2O)
bp
1560 °C/1013 hPa
mp
963 °C
solubility
water: 375 g/L at 20 °C
density
3.9 g/cm3 at 20 °C
bulk density
1350 kg/m3
storage temp.
2-30°C
SMILES string
[Ba+2].[Cl-].[Cl-]
InChI
1S/Ba.2ClH/h;2*1H/q+2;;/p-2
InChI key
WDIHJSXYQDMJHN-UHFFFAOYSA-L
Application
- Structural Characterization of Sulfated Polysaccharide Isolated From Red Algae (Gelidium crinale) and Antioxidant and Anti-Inflammatory Effects in Macrophage Cells.: This research involves the use of barium chloride in the structural characterization of a sulfated polysaccharide from red algae. The study explores its antioxidant and anti-inflammatory properties, providing insights into its potential applications in biomedical and pharmaceutical fields (Pei et al., 2021).
Analysis Note
Assay (complexometric): ≥ 99.5 %
Al (Aluminium): ≤ 0.05 ppm
Ca (Calcium): ≤ 2.00 ppm
Cd (Cadmium): ≤ 0.005 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.010 ppm
Cu (Copper): ≤ 0.005 ppm
Eu (Europium): ≤ 0.010 ppm
Fe (Iron): ≤ 0.02 ppm
K (Potassium): ≤ 5.0 ppm
La (Lanthanum): ≤ 0.010 ppm
Li (Lithium): ≤ 1.0 ppm
Mg (Magnesium): ≤ 1.00 ppm
Mn (Manganese): ≤ 0.010 ppm
Na (Sodium): ≤ 5 ppm
Ni (Nickel): ≤ 0.005 ppm
Pb (Lead): ≤ 0.005 ppm
Rb (Rubidium): ≤ 5.0 ppm
Sc (Scandium): ≤ 0.010 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 10.00 ppm
Tl (Thallium): ≤ 0.010 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.005 ppm
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form solid | form solid | form powder and chunks |
| assay 99.995% | assay ≥99.0% (complexometric) | assay ≥99.0% (complexometric) | assay 99.999% trace metals basis |
| solubility water: 375 g/L at 20 °C | solubility - | solubility - | solubility - |
| Quality Level 100 | Quality Level 200 | Quality Level 300 | Quality Level 200 |
| pH 5-8 (20 °C, 50 g/L in H2O) | pH 5.2-8.0 (25 °C, 50 g/L in H2O) | pH 5.2-8.0 (25 °C, 50 g/L in H2O) | pH - |
| mp 963 °C | mp - | mp - | mp 963 °C (lit.) |
Still not finding the right product?
Explore all of our products under Barium chloride
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2
Storage Class
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



