Skip to Content
Merck
All Photos(1)

Documents

1.01599

Supelco

4-Bromoaniline

GR for analysis

Synonym(s):

4-Bromoaniline, p-Bromoaniline, p-Bromophenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrN
CAS Number:
Molecular Weight:
172.02
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-393-9

Quality Level

form

solid

potency

456 mg/kg LD50, oral (Rat)
536 mg/kg LD50, skin (Rat)

bp

230-250 °C/1013 hPa

mp

60-64 °C

bulk density

810 kg/m3

storage temp.

15-25°C

InChI

1S/C6H6BrN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2

InChI key

WDFQBORIUYODSI-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 99 % (a/a)
Identity (IR-spectrum): passes test
Melting point: 62 - 64 °C
Fe (Iron): ≤ 0.001 %
Sulfated ash: ≤ 0.1 %
Water (according to Karl Fischer): ≤ 0.1 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service