Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 L | Please contact Customer Service for Availability | CZK 12,200.00 |
About This Item
CZK 12,200.00
Quality Segment
form
liquid
analyte functional class(es)
chemical oxygen demand (COD)
technique(s)
photometry: suitable
density
1.88 g/cm3 at 20 °C
storage temp.
15-25°C
Analysis Note
Concentration (H₂SO₄): 1665 - 1735 g/l
1 of 1
This Item | ||
|---|---|---|
| technique(s) photometry: suitable | technique(s) photometry: suitable | technique(s) photometry: suitable |
| form liquid | form liquid | form liquid |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C |
| density 1.88 g/cm3 at 20 °C | density 1.84 g/cm3 at 20 °C | density 1.3 g/cm3 at 20 °C |
| Quality Level 100 | Quality Level 100 | Quality Level 100 |
| analyte functional class(es) chemical oxygen demand (COD) | analyte functional class(es) chemical oxygen demand (COD) | analyte functional class(es) chemical oxygen demand (COD) |
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class
8B - Non-combustible, corrosive hazardous materials
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


