Skip to Content
Merck

Skip To

1.01254

Potassium chloride solution

nominal 12.8 mS/cm, certified reference material for the measurement of electrolytic conductivity (c=0.1 mol/l) Certipur®

Synonym(s):

Potassium chloride solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL
Please contact Customer Service for Availability
CZK 6,200.00

About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
1711999
MDL number:

CZK 6,200.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

ISO (17025, 17034)

product line

Certipur®

density

1.003 g/cm3

application(s)

environmental
food and beverages
pharma/biopharma processes
water monitoring

format

single component solution

storage temp.

15-25°C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

General description

This certified reference material (CRM) is directly traceable to the corresponding primary reference material from the Physikalisch Technische Bundesanstalt (PTB), Germany. The electrolytic conductivity of this standard is determined using a conductivity meter from an accredited Merck calibration laboratory.

The details of expiry date, storage and handling, and associated uncertainty are documented in the certificate of analysis.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Used as a calibration standard in aqueous solution to determine conductivity cell constant or as a control sample.

Analysis Note

Electrical conductivity (25 °C): 12.750 - 12.950 mS/cm

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.015571.015861.01203
format

single component solution

format

single component solution

format

single component solution

format

single component solution

product line

Certipur®

product line

Certipur®

product line

Certipur®

product line

Certipur®

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

environmental
food and beverages
pharma/biopharma processes
water monitoring

application(s)

environmental
food and beverages
pharmaceutical
water monitoring

application(s)

environmental
food and beverages
pharma/biopharma processes
water monitoring

application(s)

environmental

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

agency

ISO (17025, 17034)

agency

ISO (17025, 17034)

agency

-

agency

ISO (17025, 17034)


Still not finding the right product?

Explore all of our products under Potassium chloride solution


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters