Skip to Content
Merck
All Photos(1)

Documents

07-505

Sigma-Aldrich

Anti-HDAC8 Antibody

serum, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... HDAC8(55869)

Specificity

HDAC8

Immunogen

GST fusion protein corresponding to amino acids 305-377 of human HDAC8

Application

Anti-HDAC8 Antibody is a Rabbit Polyclonal Antibody for detection of HDAC8 also known as histone deacetylase 8 & has been tested in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

routinely evaluated by immunoblot in in a nuclear extract of mouse liver

Target description

48 kDa

Physical form

30% glycerol
Antiserum

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Van den Wyngaert et al.
FEBS letters, 478(1-2), 77-83 (2000-08-03)
To date, seven different human histone deacetylases (HDACs) have been identified, which fall into two distinct classes. We have isolated and characterized a cDNA encoding a novel human HDAC, which we name HDAC8. HDAC8 shows a high degree of sequence
J J Buggy et al.
The Biochemical journal, 350 Pt 1, 199-205 (2000-08-06)
Histone deacetylases (HDACs) are a growing family of enzymes implicated in transcriptional regulation by affecting the acetylation state of core histones in the nucleus of cells. HDACs are known to have key roles in the regulation of cell proliferation [Brehm
E Hu et al.
The Journal of biological chemistry, 275(20), 15254-15264 (2000-04-05)
Histone acetylation alters chromatin state by modifying lysines on histone and plays an important role in modulating gene transcription. A dynamic balance of histone acetylation/deacetylation is maintained by histone acetyltransferases and histone deacetylases. Emerging evidence suggests that a family of
Dawn Sijin Nin et al.
Cell reports, 36(9), 109621-109621 (2021-09-02)
Radiotherapy (RT) resistance is a major cause of treatment failure in cancers that use definitive RT as their primary treatment modality. This study identifies the cancer/testis (CT) antigen G antigen (GAGE) as a mediator of radio resistance in cervical cancers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service