Skip to Content

07-2270

Anti-WNK4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Serine/threonine-protein kinase WNK4, Protein kinase lysine-deficient 4, Protein kinase with no lysine 4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WNK4(65266)

General description

WNK4 is a serine-threonine protein kinase that is commonly expressed in the distal nephron. Mutations in WNK1 and WNK4 cause pseudohypoaldosteronism type II, which causes hypertension, hyperkalemia, hyperchloremia and metabolic acidosis. WNK4 has been shown to decrease Na-Cl cotransporter activity and the potassium channel by removing it from the cell surface. It is thought that activating WNK4 inhibits the activity of WNK1. WNK4 has been shown to play a role in regulating blood pressure in addition to potassium homeostasis.
~160 kDa observed. This protein has 3 isoforms produced by alternative splicing. Hengl, T., et al. (2010). PNAS. 107(13):6052–6057.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to human WNK4 at the C-terminus.

Application

Immunohistochemistry Analysis: 1:1,000 dilution from a previous lot detected WNK4 in normal human kidney tissue.
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases
This WNK4 antibody is validated for use in WB & IHC for the detection of the WNK4 protein.

Biochem/physiol Actions

This antibody recognizes WNK4 at the C-terminus.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse kidney cell lysate
Evaluated by Western Blot in mouse kidney cell lysate.

Western Blot Analysis: 0.05 µg/mL of this antibody detected WNK4 on 10 µg of mouse kidney cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library