Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
WB, cell based assay
Citations:
-
biological source
rabbit
Quality Segment
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
mouse (based on 100% sequence homology), rat (based on 100% sequence homology)
technique(s)
cell based assay: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
acetylation (Lys430)
Gene Information
human ... SRC(6714)
General description
Src is a proto-oncogenic tyrosine kinase belonging to the Src family of non-receptor tyrosine kinases. This proto-oncogene plays a pivotal role cellular processes such as proliferation, migration, transformation and regulation of embyronic development and cell growth. Src tyrosine kinases transmit integrin-dependent signals for cell movement and proliferation. The study of Src proteins have contributed to the understanding of cancer caused by abnormal cellular signaling.
~69 kDa observed.
Endogenous Src has a molecular weight of ~60 kDa; however, the myc tag adds ~9 kDa to the weight of the protein.
Endogenous Src has a molecular weight of ~60 kDa; however, the myc tag adds ~9 kDa to the weight of the protein.
Immunogen
Epitope: Acetylated Lys430
KLH-conjugated linear peptide corresponding to human Src acetylated at Lys430.
Application
Research Category
Cell Structure
Cell Structure
Research Sub Category
Cytoskeleton
Cytoskeleton
This Anti-acetylated-Src (Lys430) Antibody is validated for use in WB, Cell Function Assay for the detection of acetylated-Src (Lys430).
Western Blot Analysis: 5 µg/mL from a representative lot detected Src on 10 µg of HEK293T cells transfected with myc-tagged Src and CBP.
Biochem/physiol Actions
This antibody recognizes Src acetylated at Lys430.
Physical form
Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Western Blot/Peptide Inhibition Assay Analysis: This antibody peptide blocked on HEK293T cells transfected with myc-tagged Src and CBP.
Control
HEK293T cells transfected with myc-tagged Src and CBP
HEK293T cells transfected with myc-tagged Src and CBP
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.