Skip to Content
Merck

Skip To

07-1213

Anti-IP3 Receptor 1 Antibody

serum, from rabbit

Synonym(s):

IP3 receptor isoform 1, Type 1 InsP3 receptor, Type 1 inositol 1,4,5-trisphosphate receptor, inositol 1,4,5-triphosphate receptor, type 1, spinocerebellar ataxia 15, spinocerebellar ataxia 16

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship onJune 08, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 10,500.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
5

CZK 10,500.00


Estimated to ship onJune 08, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

canine, rat

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

dog ... Itpr1(476548)
human ... ITPR1(3708)
mouse ... Itpr1(16438)
rat ... Itpr1(25262)

General description

Approx. 300 kDa
IP3 Receptor 1 is an intracellular channel that when stimulated by inositol 1,4,5-triphosphate mediates the release of calcium from the endoplasmic reticulum. Defects within the gene ITPR1 cause a group of cerebellar disorders known as spinocerebellar ataxia type 15 (SCA15). This receptor is widely expressed in tissue.

Immunogen

Synthetic peptide from the N-terminal of human IP3 Receptor 1.

Application

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected IP3 Receptor 1 in mouse cerebellum tissue.
Research Category
Signaling
Research Sub Category
Lipid Signaling
This Anti-IP3 Receptor 1 Antibody is validated for use in IH(P), WB for the detection of IP3 Receptor 1.

Biochem/physiol Actions

The antibody reacts with the 300 kDa IP3 Receptor 1 protein.

Physical form

Rabbit serum. Liquid
Unpurified rabbit serum

Preparation Note

Maintain at -20°C in undiluted aliquots for up to 1 year date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
C2C12 cell lysates
Evaluated by Western Blot in C2C12 cell lysates.
Western Blot Analysis: 1:500 dilution of this lot detected IP3 RECPT 1 on 10 ug of C2C12 lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: AB9072

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB9076AB5882AB9074
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

mouse, human

species reactivity

mouse, rat, human

species reactivity

rat

species reactivity

human, mouse

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

serum

antibody form

serum

antibody form

affinity purified immunoglobulin

antibody form

serum

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

Explore all of our products under Anti-IP3 Receptor 1 Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters