Skip to Content
Merck
All Photos(1)

Documents

06-872

Sigma-Aldrich

Anti-IRAK Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Clec1B(56760)

Specificity

Recognizes IRAK.

Immunogen

Peptide (DRQGPEESDEFQS) corresponding to amino acids 700-712 of human IRAK

Application

Detect IRAK using this Anti-IRAK Antibody validated for use in IP & WB.
Research Category
Signaling
Research Sub Category
Immunological Signaling

Quality

routinely evaluated by immunoblot on HeLa nuclear extract

Target description

80 kDa

Physical form

Ammonium sulfate precipitation and DEAE-cellulose chromatography
Format: Purified
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Candida albicans triggers activation of distinct signaling pathways to establish a proinflammatory gene expression program in primary human endothelial cells.
Verena Muller, Dorothee Viemann, Marc Schmidt, Nicole Endres, Stephan Ludwig et al.
Journal of immunology (Baltimore, Md. : 1950) (1950)
M Trofimova et al.
The Journal of biological chemistry, 271(30), 17609-17612 (1996-07-26)
The NF-kappaB/c-Rel proteins are a family of evolutionarily conserved transcription factors activated during development that in the adult, mediate many processes including the immune response. A high degree of sequence similarity is shared between the NF-kappaB/c-Rel family of transcription factors
Bruns A Watts et al.
American journal of physiology. Renal physiology, 317(3), F705-F719 (2019-06-27)
LPS inhibits HCO3- absorption in the medullary thick ascending limb (MTAL) through a Toll-like receptor 4 (TLR4)-myeloid differentiation factor 88 (MyD88)-extracellular signal-regulated kinase (ERK) pathway that is upregulated by sepsis. Pretreatment with the nontoxic immunomodulator monophosphoryl lipid A (MPLA) prevents
Neutrophil signaling pathways activated by bacterial DNA stimulation.
Maria E Alvarez, Juan I Fuxman Bass, Jorge R Geffner, Paula X Fernandez Calotti et al.
Journal of immunology (Baltimore, Md. : 1950) (1950)
L Li et al.
The Journal of biological chemistry, 275(30), 23340-23345 (2000-05-16)
Interleukin-1 receptor-associated kinase (IRAK), a signal transducer for interleukin-1, has also been suggested to participate in the Toll-like receptor-mediated innate immune response to bacterial endotoxin lipopolysaccharide (LPS). Using the human promonocytic THP-1 cell line, we demonstrated that the endogenous IRAK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service