Skip to Content
Merck

Skip To

06-1064

Anti-ITK/EMT/TSK Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

IL2-inducible T-cell kinase, Kinase EMT, T-cell-specific kinase, Tyrosine-protein kinase Lyk, homolog of mouse T-cell itk/tsk, tyrosine-protein kinase ITK/TSK

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
-


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITK(3702)

General description

Tyrosine-protein kinase ITK/TSK (UniProt: Q08881; also known as EC:2.7.10.2, Interleukin-2-inducible T-cell kinase, IL-2-inducible T-cell kinase, Kinase EMT, T-cell-specific kinase, Tyrosine-protein kinase Lyk) is encoded by the ITK (also known as EMT, LYK) gene (Gene ID: 3702) in human. ITK/TSK is a protein tyrosine kinase that plays an essential role in regulation of the adaptive immune response. It regulates the development, function, and differentiation of conventional T-cells and non-conventional natural killer T-cells. It contains four zinc binding sites. It s N-terminal PH domain allows it to be recruited to the plasma membrane by an activated PI3 kinase. This domain also contains a proline-rich region (PRR). The adjoining SH3 domain binds to PRR. The SH2 domain is required for binding tyrosine-phosphorylated substrates and the kinase domain located in the C-terminal region is required for tyrosine phosphorylation. ITK/TSK is phosphorylated at Tyr512 in the activation loop of the kinase domain by LCK and subsequent autophosphorylation at Tyr180, which lies within the substrate-binding sequence in the SH3 domain, allows the kinase activation. Mutations in ITK gene are reported to cause lymphoproliferative syndrome 1, a rare immunodeficiency characterized by extreme susceptibility to infection with Epstein-Barr virus that can lead to fatal outcome.
~71 kDa observed; 71.83 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

GST-tagged recombinant fragment cvorresponding to the first 270 amino acids from the N-terminal half of human Tyrosine-protein kinase ITK/TSK spanning the PH, SH3 and a small portion of the SH2 domains.

Application

Immunoprecipitation Analysis: A 1:500 dilution from a representative lot immunoprecipitated ITK/EMT/TSK in 500 µg of Jurkat cell lysate.
Research Category
Signaling

Inflammation & Immunology
Research Sub Category
Immunological Signaling
This rabbit polyclonal Anti-ITK/EMT/TSK, Cat. No. 06-1064 is a rabbit polyclonal antibody that targets protein tyrosine kinase ITK/TSK and is tested for use in Immunoprecipitation and Western blotting.

Biochem/physiol Actions

Predicted to cross-react with mouse based on 100% sequence homology.
Recognizes Itk/Emt/Tsk, Mr ~72 kDa.

Physical form

Affinity purified
Purified rabbit polycloncal in buffer containing PBS with 0.05% sodium azide and 50% glycerol.

Preparation Note

Stable for 1 year at -20°C from date of receipt. For maximum recovery of the product, centrifuge the original vial and prior to removing the cap. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol-containing solutions to become frozen during storage.

Analysis Note

Control
Jurkat cell lysates
Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected ITK/EMT/TSK in 10 µg of Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 06-546

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABF13104-1578FCMAB404PE
biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

mouse

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

conjugate

PE

clone

polyclonal

clone

polyclonal

clone

10D11, monoclonal

clone

Ab33, monoclonal

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified antibody

species reactivity

human

species reactivity

human

species reactivity

rat, human

species reactivity

human

UniProt accession no.

Q08881

UniProt accession no.

Q02763

UniProt accession no.

Q06187

UniProt accession no.

Q02763


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters